missing translation for 'onlineSavingsMsg'
Learn More

Sodium methanesulfonate, 99%, Thermo Scientific Chemicals

Catalog Number p-7077281
Click to view available options
Quantity:
5 g
100 g
500 g
This item is not returnable. View return policy
2386-57-4
CH4NaO3S
118.09
KKVTYAVXTDIPAP-UHFFFAOYSA-M
sodium methanesulfonate, methanesulfonic acid sodium salt, methanesulfonic acid, sodium salt, sodium methanesulphonate, sodium mesylate, methylsulfonic acid sodium salt, 75-75-2 parent, sodium methyl sulfonate, methanesulfonic acid, sodium salt 1:1, sodium methylsulfonate
638112
sodium;methanesulfonate
CS(=O)(=O)[O-].[Na+]
This item is not returnable. View return policy
2386-57-4
CH4NaO3S
118.09
KKVTYAVXTDIPAP-UHFFFAOYSA-M
sodium methanesulfonate, methanesulfonic acid sodium salt, methanesulfonic acid, sodium salt, sodium methanesulphonate, sodium mesylate, methylsulfonic acid sodium salt, 75-75-2 parent, sodium methyl sulfonate, methanesulfonic acid, sodium salt 1:1, sodium methylsulfonate
638112
sodium;methanesulfonate
CS(=O)(=O)[O-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Linear Formula CH3SO3Na
Solubility Information Solubility in water: soluble.
Formula Weight 118.09
Percent Purity 99%
Infrared Spectrum Authentic
Chemical Name or Material Sodium methanesulfonate
compliance-icons
Product Identifier
  • Sodium methanesulfonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'