Få mere at vide
Sodium Methanesulfinate-d3, CDN
Artikelnummer. 30315566
Skift visning
Mængde:
0.05 g
Artikelnummer. Mængde
30315566 0.05 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315566 Leverandør CDN Leverandørnr. CDND81890.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl-d3-sulfinic acid sodium salt
CAS 2129597-38-0
Molekylær formel C D3 O2 S . Na
Mængde 0.05 g
Synonym Methane-d3-sulfinic acid, sodium salt (1:1) (ACI), Sodium methane-d3-sulfinate, Sodium methylsulfinate-d3, Methane-d3-sulfinic acid sodium salt, Sodium methanesulfinate-D3
InChI formel InChI=1 S/CH4O2S.Na/c1-4(2)3;/h1H3,(H,2,3);/q;+1/p-1/i1D3;
SMIL [Na+].[2 H]C([2 H])([2 H])S(=O)[O-]
IUPAC navn sodium;trideuteriomethanesulfinate
Molekylvægt (g/mol) 105.1066
Formel vægt 104.994 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity