Få mere at vide
Sodium Lauryl Sulfoacetate, TRC
Artikelnummer. 30011272
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011272 50 mg
30004949 100 mg
30007087 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011272 Leverandør Toronto Research Chemicals Leverandørnr. TRCS63430350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1847-58-1
Molekylær formel C14H27O5SNa
Molekylvægt (g/mol) 330.42
Synonym Dodecyl sodium sulfoacetate, Lathanol, Lathanol LAL, Lathanol LAL 70, Miconate SLSA, Nacconol LAL, Nikkol LSA, Sodium lauryl sulfoacetate, Sulfo-dodecyl Ester Acetic Acid Sodium Salt (6CI), Sulfo-dodecyl Ester Acetic Acid S-Sodium Salt (7CI), Sulfo-1-dodecyl Ester Acetic Acid Sodium Salt (8CI,9CI);
IUPAC navn sodium;2-dodecoxy-2-oxoethanesulfonate
SMIL [Na+].CCCCCCCCCCCCOC(=O)CS(=O)(=O)[O-]
Tekniske data
Kemisk navn eller materiale Sodium Lauryl Sulfoacetate
Mængde 50 mg
InChI formel InChI=1S/C14H28O5S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-19-14(15)13-20(16,17)18;/h2-13H2,1H3,(H,16,17,18);/q;+1/p-1
Formel vægt 330.1477