missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Natriumhypochlorit, 5 % w/v, Clorox , Scharlab
Description
- Synonymer: Clorox
- Molekylformel: NaOCl
- Molmasse: 74,44 g/mol
- CAS: 7681-52-9
Specifications
Specifications
| CAS | 7681-52-9 |
| Farve | Gul |
| Fysisk form | Løsning |
| Mængde | 1 L |
| Molekylær formel | ClNaO |
| Synonym | sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution |
| InChI nøgle | SUKJFIGYRHOWBL-UHFFFAOYSA-N |
| SMIL | [O-]Cl.[Na+] |
| IUPAC navn | natrium; hypochlorit |
| Molekylvægt (g/mol) | 74.439 |
| Show More |
Safety and Handling
Product Identifier
- Natrium hypochlorite, opløsning 5% w/v, extra pure
- Fare
- Alvorlige øjenskader/øjenirritation Kategori 1
- Hudætsning/hudirritation Kategori 2
- H315-Forårsager hudirritation.
- H318-Forårsager alvorlig øjenskade.
- P280-Bær beskyttelseshandsker/beskyttelsestøj/øjenbeskyttelse/ ansigtsbeskyttelse.
- P305+P351+P338-VED KONTAKT MED ØJNENE: Skyl forsigtigt med vand i flere minutter. Fjern eventuelle kontaktlinser, hvis dette kan gøres let. Fortsæt skylning.
- P310-Ring omgående til en GIFTINFORMATION/læge/.
- P321-Særlig behandling (se på denne etiket).
- P362-Alt tilsmudset tøj tages af.
- P332+P313-Ved hudirritation: Søg lægehjælp.
- MIXTURE LIST-Contain: Sodium hypochlorite
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?