Learn More
Sodium hypochlorite, 15% w/v in water, Scharlab
Product Code. 12495122
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Product Code. Quantity Unit Size
12495122 1 L 1 L
1 options
Product Code. 12495122 Supplier Scharlab Supplier No. SO04361000
 Restricted Item
Forventet forsendelse: 15-12-2026
Specifications
CAS 7681-52-9
Quantity 1 L
Molecular Formula ClNaO
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
SMILES [Na+].[O-]Cl
IUPAC Name sodium hypochlorite
Molecular Weight (g/mol) 74.44
Grade EssentQ
compliance-icons
Product Identifier
  • Sodium hypochlorite, solution 15% w/v, extra pure
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: Sodium Hypochlorite,solution