missing translation for 'onlineSavingsMsg'
Learn More
Sodium hydroxide, 0.01N Standardized Solution Product Code.: 11323737

Sodium hydroxide, 0.01N Standardized Solution

Product Code. p-7050109
1 L, 1 L
Click to view available options
Quantity:
1 L
4 L
20 L
Unit Size:
1 L
20 L
4 L
This item is not returnable. View return policy

Product Code. 11323737

Brand: Thermo Scientific Alfa Aesar 035620.K2

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1310-73-2 | HNaO | 39.997 g/mol

Sodium hydroxide, 0.01N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Concentration 0.01N
Solubility Information Miscible with glycerol,ethanol and methanol. Immiscible with acetone and ether.
Formula Weight 40
Odor Odorless
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Sodium hydroxide, Standardized Solution

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number62500237 Certificate TypeCertificate of Analysis Date15/07/2025 Product Code035620.M4,035620.K2,035620.K7
Lot NumberP19L520 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code035620.M4,035620.K2,035620.K7
Lot NumberR01L544 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code035620.M4,035620.K2,035620.K7
Lot Number62500135 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code035620.M4,035620.K2,035620.K7
Lot NumberP16L501 Certificate TypeCertificate of Analysis Date21/03/2025 Product Code035620.M4,035620.K2,035620.K7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium hydroxide, 0.01N Standardized Solution >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.