missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydrosulfite, Honeywell Fluka™
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
2 kg
Specifications
Quantity | 100 g |
Linear Formula | NaO2SSO2Na |
UN Number | 1384 |
Solubility Information | Soluble in water |
Formula Weight | 174.11g/mol |
Grade | Technical |
Concentration or Composition (by Analyte or Components) | 85% |
Chemical Name or Material | Sodium Hydrosulfite |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'