Learn More
Sodium hydrogen L-tartrate, anhydrous, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Sodium hydrogen L-tartrate is used for detecting potassium and in nutrient media. It also used as a food additive and an acidity regulator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium hydrogen L-tartrate is used for detecting potassium and in nutrient media. It also used as a food additive and an acidity regulator.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Sodium hydrogen L-tartrate is used for detecting potassium and in nutrient media. It also used as a food additive and an acidity regulator.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | ∽243°C (decomposition) |
Quantity | 25 g |
Beilstein | 6099125 |
Merck Index | 14,8589 |
Solubility Information | Soluble in water. |
Optical Rotation | +24.5° (c=3 in Water) |
Formula Weight | 172.07 |
Percent Purity | 98% |
Chemical Name or Material | Sodium hydrogen L-tartrate, anhydrous |