Få mere at vide
Sodium Formate-d, TRC
Artikelnummer. 30418434
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30418434 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418434 Leverandør Toronto Research Chemicals Leverandørnr. TRCS629002100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sodium deuterioformate
CAS 3996-15-4
Molekylær formel C 2H O2 . Na
Mængde 100 mg
Synonym Formic-d acid','sodium salt (8CI','9CI), Sodium formate-d (6CI), Sodium deuterioformate, Sodium formate-d1, Sodium formate-D
InChI formel InChI=1S/CH2O2.Na/c2-1-3;/h1H,(H,2,3);/q;+1/p-1/i1D;
SMIL [Na+].[2H]C(=O)[O-]
IUPAC navn sodium;deuterioformate
Molekylvægt (g/mol) 69.01
Formel vægt 68.9937