missing translation for 'onlineSavingsMsg'
Learn More

Sodium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7080248
Click to view available options
Quantity:
10 g
50 g
7681-49-4
FNa
41.99
MFCD00003524
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
5235
CHEBI:28741
sodium fluoride
[F-].[Na+]
7681-49-4
FNa
41.99
MFCD00003524
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
5235
CHEBI:28741
sodium fluoride
[F-].[Na+]

Sodium fluoride is used in laundry souring, manufacturing enamels, manufacturing coated papers, wood preservative , foam production, opal glass, ore flotation and water fluoridation. It is also acts as an inhibitor of serine/threonine phosphatases widely used in bone and dental research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1704°C
Melting Point 993°C
Color White
Physical Form Pieces
Quantity 10 g
Linear Formula NaF
UN Number UN1690
Merck Index 14,8618
Solubility Information It is soluble in water,slightly soluble in HF,ammonia. Negligible in alcohol,acetone,SO2,dimethylformamide.
Formula Weight 41.99
Percent Purity 99.99%
Odor Odorless
Refractive Index 1.336
Density 2.78 g/mL
Assay (metals basis)
Chemical Name or Material Sodium fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'