Få mere at vide
Sodium Ethylxanthate (~90%), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Sodium Ethylxanthate |
| CAS | 140-90-9 |
| Molekylær formel | C3H5NaOS2 |
| Mængde | 250 g |
| Synonym | O-Ethyl Ester Dithio-carbonic Acid','Sodium Salt, O-Ethyl Ester Carbonodithioic Acid','Sodium Salt, Ethyl-xanthic Acid','Sodium Salt, Aero 325, Aeroxanthate 325, Ethylxanthic Acid Sodium Salt, O-Ethyl Sodium Dithiocarbonate, SEX, SEX (flotation agent), Sodium O-Ethyl Dithiocarbonate, Sodium O-Ethyl Carbondoithioate, Sodium O-Ethyl Xanthate, Sodium Ethyl Xanthogenate, Sodium Xanthate, Sodium Xanthogenate, Z 4, Z 4 (flotation agent) |
| InChI formel | InChI=1S/C3H6OS2.Na/c1-2-4-3(5)6;/h2H2,1H3,(H,5,6);/q;+1/p-1 |
| SMIL | [Na+].CCOC(=S)[S-] |
| IUPAC navn | sodium;ethoxymethanedithioate |
| Molekylvægt (g/mol) | 144.19 |
| Formel vægt | 143.968 |