Få mere at vide
Sodium Ethyl Parahydroxybenzoate, Mikromol™
Beskrivelse
Sodium Ethyl Parahydroxybenzoate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Sodium Ethyl Parahydroxybenzoate |
| CAS | 35285-68-8 |
| Analyt- eller komponentnavne | Sodium Ethyl Parahydroxybenzoate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C9 H9 O3 . Na |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Sodium Ethyl Parahydroxybenzoate, p-Hydroxybenzoic acid ethyl ester sodium deriv., 4-Hydroxybenzoic acid ethyl ester sodium salt, Ethyl 4-hydroxybenzoate sodium salt, Ethyl p-hydroxybenzoate sodium salt, Sodium ethyl p-hydroxybenzoate, Sodium p-(carbethoxy)phenoxide |
| InChI formel | InChI=1S/C9H10O3.Na/c1-2-12-9(11)7-3-5-8(10)6-4-7;/h3-6,10H,2H2,1H3;/q;+1/p-1 |
| SMIL | [Na+].CCOC(=O)c1ccc([O-])cc1 |
| IUPAC navn | sodium;4-ethoxycarbonylphenolate |
| Vis mere |