Få mere at vide
Sodium Dodecyl Sulfate-d25, TRC
Artikelnummer. 30425517
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30425517 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30425517 Leverandør Toronto Research Chemicals Leverandørnr. TRCS083920100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sodium Dodecyl Sulfate-d25
CAS 110863-24-6
Renhedsgrad noter HPLC
Molekylær formel C12 D25 O4 S . Na
Mængde 100 mg
Synonym 1-Dodecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-d25-ol','hydrogen sulfate','sodium salt (1:1) (ACI), 1-Dodecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-d25-ol','hydrogen sulfate','sodium salt (9CI), Sodium d25-dodecyl sulfate, Sodium dodecyl sulfate-d25, Dodecyl-D25 sodium sulfate, Perdeuterododecyl sulfate sodium salt, Sodium lauryl sulfate-d25, Laurylsulfate-d25 sodium
InChI formel InChI=1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2;
SMIL [Na+].[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])OS(=O)(=O)[O-]
IUPAC navn sodium;1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-pentacosadeuteriododecyl sulfate
Molekylvægt (g/mol) 313.533
Formel vægt 313.294
Procent renhed >95