missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Sodium dodecamolybdophosphate hydrate, 90+%
Click to view available options
Quantity:
100 g
500 g
Specifications
Quantity | 100 g |
Merck Index | 14,8664 |
Formula Weight | 1891.22 (Anhydrous) |
Percent Purity | ≥90% |
Chemical Name or Material | Sodium dodecamolybdophosphate hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'