Få mere at vide
Sodium DL-Lactate-2,3,3,3-d4 (60% w/w in H2O), TRC
Artikelnummer. 30419967
Skift visning
Mængde:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30419967 10 mg
30421071 1 mg
30416899 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419967 Leverandør Toronto Research Chemicals Leverandørnr. TRCS64400410MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 344299-52-1
Molekylær formel C3 D4 H O3 . Na
Molekylvægt (g/mol) 116.08
Synonym Propanoic-2,3,3,3-d4 acid, 2-hydroxy-, monosodium salt (9CI)
IUPAC navn sodium;2,3,3,3-tetradeuterio-2-hydroxypropanoate
SMIL [Na+].[2H]C([2H])([2H])C([2H])(O)C(=O)[O-]
Tekniske data
Kemisk navn eller materiale Sodium DL-Lactate-2,3,3,3-d4
Mængde 10 mg
InChI formel InChI=1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/i1D3,2D;
Formel vægt 116.0387
Koncentration eller sammensætning (efter analyt eller komponenter) 60%