Få mere at vide
Sodium DL-Lactate-2,3,3,3-d4 (60% w/w in H2O), CDN
Artikelnummer. 30317783
Skift visning
Mængde:
0.1 g
Pakningsstørrelse:
0.1g
Artikelnummer. Mængde unitSize
30317783 0.1 g 0.1g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317783 Leverandør CDN Leverandørnr. CDND22730.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sodium DL-Lactate-2,3,3,3-d4
CAS 344299-52-1
Opløsningsmiddel eller Matrix H2O
Molekylær formel C3 D4 H O3 . Na
Mængde 0.1 g
Synonym Propanoic-2,3,3,3-d4 acid, 2-hydroxy-, monosodium salt (9 CI)
InChI formel InChI=1 S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4 H,1H3,(H,5,6);/q;+1/p-1/i1D3,2 D;
SMIL [Na+].[2 H]C([2 H])([2 H])C([2 H])(O)C(=O)[O-]
IUPAC navn sodium;2,3,3,3-tetradeuterio-2-hydroxypropanoate
Molekylvægt (g/mol) 116.08
Formel vægt 116.0387 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity
Koncentration eller sammensætning (efter analyt eller komponenter) 60 %