Learn More
Sodium Dihydrogen Phosphate Monohydrate
Product Code. 15864800
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
Product Code. Quantity Packaging Unit Size
15864800 1 kg Plastic Bottle 1kg
1 options
Product Code. 15864800 Supplier Supelco Supplier No. 1063461000
Supelco Essentials Are in Stock
Use promo code 27958
View all promotions
 Restricted Item
På lager
Add to basket
Edge
Add to basket
Chemical Identifiers
CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
IUPAC Name sodium phosphoric acid hydrate
SMILES O.[Na+].OP(O)(O)=O
Specifications
Physical Form Crystals
Quantity 1 kg
Formula Weight 137.99
Percent Purity 99.0 to 102.0%
Grade ACS
Packaging Plastic Bottle
Density 2.04 g/cm3 (20°C); Bulk Density: 880 kg/m3
Chemical Name or Material Sodium Dihydrogen Phosphate Monohydrate