missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride (colorless to white crystalline granules/USP/FCC/EP/BP/JP), Fisher Chemical™

Catalog Number 12357369
Encompass
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
7647-14-5
ClNa
58.44
MFCD00003477
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
5234
CHEBI:26710
sodium chloride
[Na+].[Cl-]
This item is not returnable. View return policy
7647-14-5
ClNa
58.44
MFCD00003477
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
5234
CHEBI:26710
sodium chloride
[Na+].[Cl-]

This product is not intended for dialysis, parenteral or injectable use without further processing

Boiling Point 1461°C
Melting Point 801°C
Color White
pH 5.0 to 8.0
Physical Form Solid
Quantity 500 g
Formula Weight 58.44
Percent Purity 99.0 to 100.5% (BP)
Grade BP/EP/FCC/JP/USP
Identification Pass Test (BP, SP, FCC, JP, USP), Pass Test (BP, SP, FCC, Pass Test (EP), Pass Test (FCC), Pass Test (JP), Pass Test (USP)
Loss on Drying ≤0.5% (at 110°C, FCC), ≤0.5% (BP/BP, EP/EP, JP/JP, USP/USP), 0.5% max. (BP), 0.5% max. (EP), 0.5% max. (FCC), 0.5% max. (JP), 0.5% max. (USP)
Packaging Poly Bottle
Vapor Pressure 1mmHg at 440°C
Density 2.165g/cm³
Chemical Name or Material Sodium Chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'