missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride, 99.0 to 100.5%, USP/FCC, Granular, J.T. Baker™
Click to view available options
Quantity:
4 Ea.
Unit Size:
2.50kg
Specifications
Specifications
CAS | 7647-14-5 |
Physical Form | Solid |
Quantity | 4 Ea. |
Assay Percent Range | 99.0 to 100.5% |
Molecular Formula | ClNa |
MDL Number | MFCD00003477 |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
SMILES | [Na+].[Cl-] |
IUPAC Name | sodium chloride |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Chloride, 99.0 to 100.5%, USP/FCC, Granular, J.T. Baker™ >
Spot an opportunity for improvement?Share a Content Correction