missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium carbonate, Puriss., meets analytical spec. of BP, Ph. Eur., NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance), Honeywell Fluka™
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
25 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Physical Form | Anhydrous |
Quantity | 1 kg |
Linear Formula | Na2CO3 |
UN Number | NONH for all modes of transport |
Beilstein | 4154566 |
IUPAC Name | disodium;carbonate |
Formula Weight | 105.99g/mol |
Percent Purity | 99.5 to 100.5% (calc. to the dried substance) |
Grade | puriss. |
Chemical Name or Material | Sodium Carbonate, Meets analytical specification of Ph.Eur., BP, NF, FCC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'