missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium carbonate, Powder, ≥99.5%, ACS reagent, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
12 kg
Packaging:
Fiberboard Box
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Melting Point | 851°C (literature) |
CAS Min % | ≥90.0000% |
CAS Max % | ≤100.0000% |
Physical Form | Powder |
Quantity | 500 g |
Linear Formula | Na2CO3 |
Beilstein | 4154566 |
IUPAC Name | disodium;carbonate |
Formula Weight | 105.99 |
Percent Purity | >99.5% |
Show More |
Safety and Handling
Product Identifier
- sodium carbonate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : sodium carbonate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'