missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate Monohydrate, ≥99.5%, ACS Reagent, Honeywell™ Fluka™

Catalog Number p-7120776
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
5968-11-6
CH2Na2O4
124.00
MFCD00149177
MQRJBSHKWOFOGF-UHFFFAOYSA-L
sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop
2735133
disodium;carbonate;hydrate
C(=O)([O-])[O-].O.[Na+].[Na+]
This item is not returnable. View return policy
5968-11-6
CH2Na2O4
124.00
MFCD00149177
MQRJBSHKWOFOGF-UHFFFAOYSA-L
sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop
2735133
disodium;carbonate;hydrate
C(=O)([O-])[O-].O.[Na+].[Na+]

ACS Reagent,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 851°C
Color White
Quantity 100 g
Linear Formula Na2CO3 · H2O
UN Number NONH for all modes of transport
Formula Weight 124g/mol
Percent Purity ≥99.5%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder
Density 2.25g/cm3
Chemical Name or Material Sodium Carbonate Monohydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'