missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog Number p-4258716
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
497-19-8
CNa2O3
105.99
MFCD00003494
CDBYLPFSWZWCQE-UHFFFAOYSA-L
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
10340
CHEBI:29377
[Na+].[Na+].[O-]C([O-])=O
This item is not returnable. View return policy
497-19-8
CNa2O3
105.99
MFCD00003494
CDBYLPFSWZWCQE-UHFFFAOYSA-L
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
10340
CHEBI:29377
[Na+].[Na+].[O-]C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY
Boiling Point 1600.0°C
Melting Point 851.0°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 99.999%
Linear Formula Na2CO3
Merck Index 15,8731
Solubility Information Solubility in water: 22 g/100mL (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
IUPAC Name disodium carbonate
Formula Weight 105.99
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Sodium carbonate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'