Få mere at vide
Sodium Butyrate-d7, TRC
Artikelnummer. 30413463
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30413463 10 mg
30411769 50 mg
30409308 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30413463 Leverandør Toronto Research Chemicals Leverandørnr. TRCS61517910MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 270564-49-3
Molekylær formel C4D7NaO2
Molekylvægt (g/mol) 117.13
Synonym Sodium Butanoate-2,2,3,3,4,4,4-d7 (1:1)
IUPAC navn sodium butanoate-d7
SMIL O=C(C([2H])(C([2H])(C([2H])([2H])[2H])[2H])[2H])O[Na]
Tekniske data
Kemisk navn eller materiale Sodium Butanoate-D7
Mængde 10 mg
InChI formel InChI=1S/C4H8O2.Na/c1-2-3-4(5)6;/h2-3H2,1H3,(H,5,6);/q;+1/p-1/i1D3,2D2,3D2;
Formel vægt 117.08