missing translation for 'onlineSavingsMsg'
Learn More
Sodium Bromide, Honeywell Fluka™ Product Code.: 15610570

Sodium Bromide, Honeywell Fluka™

Product Code. p-7071696 Shop All Honeywell Fluka Products
500 g, 500g
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 1 kg
Unit Size:
1kg
2.50kg
500g
Pack of 6
This item is not returnable. View return policy

Product Code. 15610570

Brand: Honeywell-Fluka 02119500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
SMILES [Na+].[Br-]

Specifications

Quantity 500 g
Linear Formula NaBr
UN Number NONH for all modes of transport
IUPAC Name sodium bromide
Formula Weight 102.89g/mol
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Packaging Plastic Bottle
Vapor Pressure 1mmHg (806°C)
Chemical Name or Material Sodium Bromide, Meets analytical specification of Ph. Eur.BP,USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Bromide, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.