Learn More
Medchem Express 10mM * 1 mL NE 10790
Brand:  Medchem Express HY-16011_1mL
Encompass_Preferred
10mM * 1 mL NE 10790
Product Code. 18613333
  • Ring for pris / 1ml
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.