Learn More
Sodium bromide, 99+% (dry wt.), water <1.0%
Description
Sodium bromide is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium bromide is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.
Solubility
Soluble in water, soluble in alcohol.
Notes
It is thermally and chemically stable. Hygroscopic. Sodium bromide is non-flammable and when strongly heated it may emit bromine fumes.
Specifications
Specifications
Boiling Point | 1390°C |
Melting Point | 755°C |
Quantity | 250 g |
Merck Index | 14,8594 |
Solubility Information | Soluble in water,soluble in alcohol. |
IUPAC Name | sodium bromide |
Formula Weight | 102.89 |
Percent Purity | ≥99% |
Refractive Index | 1.6412 |
Sensitivity | Hygroscopic |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
62500264 | Certificate of Analysis | 17/05/2025 | A10552.30,A10552.0B,A10552.0I |
10251754 | Certificate of Analysis | 30/08/2024 | A10552.30,A10552.0B,A10552.0I |
10243682 | Certificate of Analysis | 13/10/2022 | A10552.30,A10552.0B,A10552.0I |
10187635 | Certificate of Analysis | 28/04/2022 | A10552.30,A10552.0B,A10552.0I |
10172570 | Certificate of Analysis | 22/11/2021 | A10552.30,A10552.0B,A10552.0I |
Safety and Handling


- Sodium bromide
- Warning
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H336-May cause drowsiness or dizziness.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Sodium bromide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.