missing translation for 'onlineSavingsMsg'
Få mere at vide

Sodium bicarbonate, 99.5%, MP Biomedicals™

Artikelnummer. 11405442 Shop alle MP Biomedicals produkter
Klik for at se tilgængelige muligheder
Mængde:
500 g
1 kg
3 kg
5 kg

Artikelnummer. 11405442

Brand: MP Biomedicals 02195497.5

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Sodium bicarbonate ≥99.5%, White Powder

  • It is broadly used as a physiological buffer in cell culture applications.
  • The pKa for bicarbonate <-> carbonic acid reaction is 6.4 The pKa for bicarbonate <-> carbonate reaction is 10.3.
  • It has two pKas, 6.4 and 10.3. The compound can react with both acids and bases, therefore it can be used as a neutralizer.
  • It may be used to remove or wash acidic impurities from a crude liquid to yield a purer sample.
  • it reacts with the carboxyl groups to give a fast-forming and effervescent CO2 formation which has been used to test for the presence of carboxylic groups in proteins.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 144-55-8
Molekylær formel CHNaO3
Molekylvægt (g/mol) 84.01
MDL nummer MFCD00003528
InChI nøgle UIIMBOGNXHQVGW-UHFFFAOYSA-M
IUPAC navn natriumhydrogencarbonat
SMIL [Na+].OC([O-])=O

Tekniske data

Smeltepunkt 270°C; decomposition begins at 50°C (literature)
Farve Hvid
pH 8.1 to 8.5 (0.1 M aq soln)
Fysisk form Pulver
Mængde 500 g
Renhedsgrad noter ≥99.5%
Beilstein 4153970
Synonym Sodium Hydrogen Carbonate, Baking Soda, Carbonic acid monosodium salt, Monosodium carbonate
Opløselighedsinformation Soluble in water (100mg/mL).
Brydningsindeks 1.500(Lit.)
Tæthed 2.159 at 20°C(Lit.)
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.