Få mere at vide
Sodium beta-Hydroxyethoxyacetate, TRC
Artikelnummer. 30409754
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30409754 100 mg
30412619 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30409754 Leverandør Toronto Research Chemicals Leverandørnr. TRCS634850100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 142047-97-0
Molekylær formel C4H7NaO4
Molekylvægt (g/mol) 142.09
Synonym (2-Hydroxyethoxy)acetic Acid Monosodium Salt, Sodium β-Hydroxyethoxyacetate
SMIL O=C(COCCO)O[Na]
Tekniske data
Kemisk navn eller materiale Sodium beta-Hydroxyethoxyacetate
Mængde 100 mg
InChI formel InChI=1S/C4H8O4.Na/c5-1-2-8-3-4(6)7;/h5H,1-3H2,(H,6,7);/q;+1/p-1
Formel vægt 142.0242