Få mere at vide
Sodium Benzoylthioethanesulfonate-d4, TRC
Artikelnummer. 30423585
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30423585 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30423585 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6145025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sodium Benzoylthioethanesulfonate-d4
CAS 1189657-00-8
Molekylær formel C9 2H4 H5 O4 S2 . Na
Mængde 5 mg
InChI formel InChI=1S/C9H10O4S2.Na/c10-9(8-4-2-1-3-5-8)14-6-7-15(11,12)13;/h1-5H,6-7H2,(H,11,12,13);/q;+1/p-1/i6D2,7D2;
SMIL [Na+].[2H]C([2H])(SC(=O)c1ccccc1)C([2H])([2H])S(=O)(=O)[O-]
IUPAC navn sodium;2-benzoylsulfanyl-1,1,2,2-tetradeuterioethanesulfonate
Molekylvægt (g/mol) 272.31
Formel vægt 272.0091