Få mere at vide
Sodium Benzoate-d5, TRC
Artikelnummer. 30416897
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30416897 10 mg
30423028 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30416897 Leverandør Toronto Research Chemicals Leverandørnr. TRCS61400110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sodium Benzoate-d5
CAS 62790-26-5
Molekylær formel C7 D5 O2 . Na
Mængde 10 mg
Synonym Benzoic-d5 acid','sodium salt (9CI), Sodium benzoate-d5, Benzoic-2,3,4,5,6-d5 acid sodium salt, 2,3,4,5,6-Pentadeuteriobenzoic acid sodium salt, Benzoic Acid-D5 (phenyl-D5) Sodium Salt
InChI formel InChI=1S/C7H6O2.Na/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);/q;+1/p-1/i1D,2D,3D,4D,5D;
SMIL [Na+].[2H]c1c([2H])c([2H])c(C(=O)[O-])c([2H])c1[2H]
IUPAC navn sodium;2,3,4,5,6-pentadeuteriobenzoate
Molekylvægt (g/mol) 149.134
Formel vægt 149.05