missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium acetate trihydrate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al, Honeywell Fluka™
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
25 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 1 kg |
Linear Formula | CH3COONa · 3H2O |
UN Number | NONH for all modes of transport |
Beilstein | 3732037 |
Formula Weight | 136.08g/mol |
Percent Purity | 99 to 101% (calc. to the dried substance) |
Grade | puriss. |
Ignition Point | 1112°F |
Chemical Name or Material | Sodium Acetate Trihydrate, Meets analytical specification of Ph.Eur.BP, USP, FCC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'