Få mere at vide
Sodium Acetate Trihydrate for analysis EMSURE™ ACS,ISO,Reag. Ph Eur, MilliporeSigma™
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Sodium acetate trihydrate |
| Smeltepunkt | 58°C |
| CAS | 6131-90-4 |
| Fysisk form | Crystalline Solid |
| Mængde | 500 g |
| Molekylær formel | C2H9NaO5 |
| Synonym | sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn |
| InChI nøgle | AYRVGWHSXIMRAB-UHFFFAOYSA-M |
| SMIL | CC(=O)[O-].O.O.O.[Na+] |
| IUPAC navn | sodium;acetate;trihydrate |
| Vis mere |