missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium acetate, Puriss., anhydrous, ≥98%, powder, Honeywell
Click to view available options
Quantity:
25 kg
Packaging:
Paper / Poly Bag
Specifications
CAS | 127-09-3 |
CAS Max % | 100.0000% |
Physical Form | Powder |
Quantity | 25 kg |
Molecular Formula | C2H3NaO2 |
Linear Formula | CH3COONa |
MDL Number | MFCD00012459 |
UN Number | NONH for all modes of transport |
Beilstein | 3595639 |
Flash Point | >250°C (>482°F) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'