missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium acetate, Free-flowing, ACS reagent, anhydrous, ≥99.0%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Melting Point | >300°C (decomposition)(literature) |
CAS Min % | ≥90.0000% |
CAS Max % | ≤100.0000% |
Physical Form | Anhydrous |
Quantity | 500 g |
Linear Formula | CH3COONa |
Beilstein | 3595639 |
Formula Weight | 82.03 |
Percent Purity | >99% |
Grade | ACS Reagent |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'