Få mere at vide
Sodium Acetate (2-13C), TRC
Artikelnummer. 30135433
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135433 10 mg
30134049 100 mg
17937254 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135433 Leverandør Toronto Research Chemicals Leverandørnr. TRCS96029010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13291-89-9
Molekylær formel C13CH3NaO2
Molekylvægt (g/mol) 83.03
Synonym Sodium [2-13C]acetate, Sodium Acetate-2-13C, [2-13C]Sodium Acetate
IUPAC navn sodium;acetate
SMIL [Na+].[13CH3]C(=O)[O-]
Tekniske data
Kemisk navn eller materiale Sodium Acetate (2-13C)
Mængde 10 mg
InChI formel InChI=1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1/i1+1;
Formel vægt 83.0064