Få mere at vide
Sodium Acetate-13C2,D3, TRC
Artikelnummer. 30423815
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30423815 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30423815 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6109722.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sodium Acetate-13C2,D3
CAS 123333-80-2
Molekylær formel 13C2D3NaO2
Mængde 2.5 mg
Synonym Acetic Acid-13C2,D3 Sodium Salt, Sodium Acetate-13C2,D3, Anhydrous Aodium Acetate-13C2,D3, Cryotech NAAC-13C2,D3, Protecta Super-13C2,D3, Safeway SD-13C2,D3, Sanmi-Ace 42-13C2,D3
InChI formel InChI=1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1/i1+1D3,2+1;
SMIL [Na+].[2H][13C]([2H])([2H])[13C](=O)[O-]
IUPAC navn sodium;2,2,2-trideuterioacetate
Molekylvægt (g/mol) 87.04
Formel vægt 87.0286