Få mere at vide
Sodium 6-Hydroxy-2-naphthalenesulfonate, Mikromol™
Artikelnummer. 30051914
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051914 100 mg
30050778 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051914 Leverandør Mikromol Leverandørnr. MM3472.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Sodium 6-Hydroxy-2-naphthalenesulfonate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Hydroxy-2-naphthalenesulfonic Acid Sodium Salt
CAS 135-76-2
Analyt- eller komponentnavne 6-Hydroxy-2-naphthalenesulfonic Acid Sodium Salt
Forsendelsestilstand Room Temperature
Molekylær formel C10 H7 O4 S . Na
Certificeringer/compliance ISO 9001
Synonym 6-Hydroxy-2-naphthalenesulfonic Acid Sodium Salt (1:1), 2-Naphthalenesulfonic acid, 6-hydroxy-, monosodium salt (8CI,9CI), 2-Hydroxy-6-naphthalenesulfonic acid sodium salt, 2-Naphthol-6-sodium sulfonate, 2-Naphthol-6-sulfonic acid sodium salt, 6-Sulfo-2-naphthol sodium salt, Schaeffer salt, Schaeffer's salt, Sodium 2-naphthol-6-sulfonate, Sodium 6-hydroxy-2-naphthalenesulfonate, Sodium beta-naphthol-6-sulfonate
InChI formel InChI=1S/C10H8O4S.Na/c11-9-3-1-8-6-10(15(12,13)14)4-2-7(8)5-9;/h1-6,11H,(H,12,13,14);/q;+1/p-1
SMIL [Na+].Oc1ccc2cc(ccc2c1)S(=O)(=O)[O-]
IUPAC navn sodium;6-hydroxynaphthalene-2-sulfonate
Molekylvægt (g/mol) 246.22
Mængde 100 mg
Formel vægt 245.9963 g/mol