Learn More
Sodium 5,5'-((2-hydroxypropane-1,3-diyl)bis(oxy))bis(4-oxo-4H-chromene-2-carboxylate), 97%, BLD Pharm™
Specifications
Specifications
| CAS | 15826-37-6 |
| Molecular Formula | C23H14Na2O11 |
| MDL Number | MFCD00057744 |
| Quantity | 10 g |
| SMILES | OC(COC1=C2C(C=C(C([O-])=O)OC2=CC=C1)=O)COC3=C4C(C=C(C([O-])=O)OC4=CC=C3)=O.[Na+].[Na+] |
| Molecular Weight (g/mol) | 512.33 |
| Percent Purity | 97% |
| Chemical Name or Material | Sodium 5,5'-((2-hydroxypropane-1,3-diyl)bis(oxy))bis(4-oxo-4H-chromene-2-carboxylate) |