Learn More
Sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulfonate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 521-24-4 |
| Molecular Formula | C10H5NaO5S |
| MDL Number | MFCD00001700 |
| Quantity | 25 g |
| SMILES | O=S(C1=CC(C(C2=C1C=CC=C2)=O)=O)([O-])=O.[Na+] |
| Molecular Weight (g/mol) | 260.20 |
| Percent Purity | 98% |
| Chemical Name or Material | Sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulfonate |