Learn More
Sodium 3-(10H-Phenothiazin-10-yl)propane-1-sulfonate, TRC
Product Code. 30421842
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity Unit Size
30421842 500 mg 500mg
1 options
Product Code. 30421842 Supplier Toronto Research Chemicals Supplier No. TRCS667015500MG
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sodium 3-(10H-Phenothiazin-10-yl)propane-1-sulfonate
CAS 101199-38-6
Molecular Formula C15H14NNaO3S2
Quantity 500 mg
Synonym 10H-Phenothiazine-10-propanesulfonic Acid','Sodium Salt, 3-(10'-Phenothiazinyl)propane-1-sulfonate
InChI Formula InChI=1S/C15H15NO3S2.Na/c17-21(18,19)11-5-10-16-12-6-1-3-8-14(12)20-15-9-4-2-7-13(15)16;/h1-4,6-9H,5,10-11H2,(H,17,18,19);/q;+1/p-1
SMILES [Na+].[O-]S(=O)(=O)CCCN1c2ccccc2Sc3ccccc13
IUPAC Name sodium;3-phenothiazin-10-ylpropane-1-sulfonate
Molecular Weight (g/mol) 343.4
Formula Weight 343.0313