Få mere at vide
Sodium 2-Methyl-4,5-dihydro-1,3-thiazol-4-carboxylate, Mikromol™
Artikelnummer. 17991666
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17991666 100 mg
17991136 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17991666 Leverandør Mikromol Leverandørnr. MM0024.05
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Sodium 2-Methyl-4,5-dihydro-1,3-thiazol-4-carboxylate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sodium 2-Methyl-4,5-dihydro-1,3-thiazol-4-carboxylate
CAS 15058-19-2
Analyt- eller komponentnavne Sodium 2-Methyl-4,5-dihydro-1,3-thiazol-4-carboxylate
Forsendelsestilstand Room Temperature
Molekylær formel C5 H6 N O2 S . Na
Certificeringer/compliance ISO 17025
Synonym Sodium 2-Methyl-4,5-dihydro-1,3-thiazol-4-carboxylate
InChI formel InChI=1S/C5H7NO2S.Na/c1-3-6-4(2-9-3)5(7)8;/h4H,2H2,1H3,(H,7,8);/q;+1/p-1
SMIL [Na+].CC1=NC(CS1)C(=O)[O-]
IUPAC navn sodium;2-methyl-4,5-dihydro-1,3-thiazole-4-carboxylate
Molekylvægt (g/mol) 167.16
Mængde 100 mg
Formel vægt 167.0017 g/mol