Få mere at vide
Sodium 2-Iodobenzenesulfonate, TRC
Artikelnummer. 30179524
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179524 100 mg
30179454 1 g
30179028 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179524 Leverandør Toronto Research Chemicals Leverandørnr. TRCS638005100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 62973-69-7
Molekylær formel C6H5IO3S.Na
Molekylvægt (g/mol) 307.06
Synonym 2-Iodo-Benzenesulfonic Acid Sodium Salt, O-Iodo-Benzenesulfonic Acid Sodium Salt;
IUPAC navn 2-iodobenzenesulfonic acid, sodium salt
SMIL [Na].O=S(=O)(O)C=1C=CC=CC1I
Tekniske data
Kemisk navn eller materiale Sodium 2-Iodobenzenesulfonate
Mængde 100 mg
InChI formel InChI=1S/C6H5IO3S.Na/c7-5-3-1-2-4-6(5)11(8,9)10;/h1-4H,(H,8,9,10);
Formel vægt 306.89