Få mere at vide
Sodium 1H,1H,2H,2H-Perfluorohexane Sulfonate, TRC
Artikelnummer. 30008512
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008512 25 mg
30007269 5 mg
30006853 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008512 Leverandør Toronto Research Chemicals Leverandørnr. TRCS67320025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 27619-93-8
Molekylær formel C6 H4 F9 O3 S . Na
Molekylvægt (g/mol) 350.13
Synonym 1-Hexanesulfonic acid, 3,3,4,4,5,5,6,6,6-nonafluoro-, sodium salt (1:1), 1-Hexanesulfonic acid, 3,3,4,4,5,5,6,6,6-nonafluoro-, sodium salt (8CI,9CI), 1H,1H,2H,2H-Perfluorohexanesulfonic acid sodium
IUPAC navn sodium;3,3,4,4,5,5,6,6,6-nonafluorohexane-1-sulfonate
SMIL [Na+].[O-]S(=O)(=O)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Tekniske data
Kemisk navn eller materiale 1H,1H,2H,2H-Perfluorohexanesulfonic acid sodium
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C6H5F9O3S.Na/c7-3(8,1-2-19(16,17)18)4(9,10)5(11,12)6(13,14)15;/h1-2H2,(H,16,17,18);/q;+1/p-1
Formel vægt 349.9635
Procent renhed >95%