Få mere at vide
Sodium 17beta-Estradiol-2,4,16,16-d4 3-Sulfate (Stabilized with TRIS, 50% w/w), TRC
Artikelnummer. 30372732
Skift visning
Mængde:
2 mg
Artikelnummer. Mængde
30372732 2 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30372732 Leverandør Toronto Research Chemicals Leverandørnr. TRCE8888262MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sodium 17beta-Estradiol-2,4,16,16-d4 3-Sulfate
CAS 352431-50-6
Molekylær formel C18 D4 H19 O5 S . Na
Mængde 2 mg
Synonym Sodium 17β-estradiol-2,4,16,16-d4 3-sulfate, (17β)-17-Hydroxyestra-1,3,5(10)-trien-3-yl-2,4,16,16-d4 hydrogen sulfate sodium salt, 2,4,16,16-Tetradeuterio-1,3,5(10)-estratriene-3,17β-diol-3-sulfate sodium salt, 17β-Estradiol-2,4,16,16-d4 3-sulfate sodium salt, Sodium 17β-Estradiol sulfate-d4, 17β-Estradiol sodium sulfate 2,4,16,16-D4
InChI formel InChI=1S/C18H24O5S.Na/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;/h3,5,10,14-17,19H,2,4,6-9H2,1H3,(H,20,21,22);/q;+1/p-1/t14-,15-,16+,17+,18+;/m1./s1/i3D,7D2,10D;
SMIL [Na+].[2H]c1cc2[C@H]3CC[C@@]4(C)[C@@H](CC([2H])([2H])[C@@H]4O)[C@@H]3CCc2c([2H])c1OS(=O)(=O)[O-]
IUPAC navn sodium;[(8R,9S,13S,14S,17S)-2,4,16,16-tetradeuterio-17-hydroxy-13-methyl-7,8,9,11,12,14,15,17-octahydro-6H-cyclopenta[a]phenanthren-3-yl] sulfate
Molekylvægt (g/mol) 378.452
Formel vægt 378.141