Få mere at vide
Sodium 17beta-Estradiol-2,4,16,16-d4 3-Sulfate (Stabilized with TRIS, 50% w/w), CDN
Artikelnummer. 30321456
Skift visning
Mængde:
0.01 g
0.02 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321456 0.01 g
30320561 0.02 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321456 Leverandør CDN Leverandørnr. CDND52870.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sodium 17 beta-Estradiol-2,4,16,16-d4 3-Sulfate
CAS 352431-50-6
Molekylær formel C18 D4 H19 O5 S . Na
Mængde 0.01 g
Synonym Sodium 17β-estradiol-2,4,16,16-d4 3-sulfate, (17β)-17-Hydroxyestra-1,3,5(10)-trien-3-yl-2,4,16,16-d4 hydrogen sulfate sodium salt, 2,4,16,16-Tetradeuterio-1,3,5(10)-estratriene-3,17β-diol-3-sulfate sodium salt, 17β-Estradiol-2,4,16,16-d4 3-sulfate sodium salt, Sodium 17β-Estradiol sulfate-d4, 17β-Estradiol sodium sulfate 2,4,16,16-D4
InChI formel InChI=1 S/C18H24O5S.Na/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;/h3,5,10,14-17,19 H,2,4,6-9H2,1H3,(H,20,21,22);/q;+1/p-1/t14-,15-,16+,17+,18+;/m1./s1/i3D,7D2,10 D;
SMIL [Na+].[2 H]c1cc2[C@H]3 CC[C@@]4(C)[C@@H](CC([2 H])([2 H])[C@@H]4 O)[C@@H]3 CCc2c([2 H])c1OS(=O)(=O)[O-]
IUPAC navn sodium;[(8 R,9 S,13 S,14 S,17 S)-2,4,16,16-tetradeuterio-17-hydroxy-13-methyl-7,8,9,11,12,14,15,17-octahydro-6 H-cyclopenta[a]phenanthren-3-yl] sulfate
Molekylvægt (g/mol) 378.452
Formel vægt 378.141 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity