Learn More
Sodium 1-undecanesulfonate, 98+% (dry wt.), water <2%, Thermo Scientific Chemicals
Catalog Number p-7025028
Click to view available options
Quantity:
5 g
25 g
100 g
5838-34-6
C20H14N4O4S
406.42
MFCD00053627
RFNALTSHRWGOHV-UHFFFAOYSA-N
sodium 1-undecanesulfonate, sodium undecane-1-sulfonate, 1-undecanesulfonic acid sodium salt, sodium undecylsulfonate, ipc-alks-11, acmc-1bn99, 1-undecanesulfonicacid, sodium salt 1:1
23692564
sodium;undecane-1-sulfonate
O=C(CN1C(=O)C2=CC=CC=C2C1=O)NC1=CC=C(C=C1)C(=O)NC1=NC=CS1
5838-34-6
C20H14N4O4S
406.42
MFCD00053627
RFNALTSHRWGOHV-UHFFFAOYSA-N
sodium 1-undecanesulfonate, sodium undecane-1-sulfonate, 1-undecanesulfonic acid sodium salt, sodium undecylsulfonate, ipc-alks-11, acmc-1bn99, 1-undecanesulfonicacid, sodium salt 1:1
23692564
sodium;undecane-1-sulfonate
O=C(CN1C(=O)C2=CC=CC=C2C1=O)NC1=CC=C(C=C1)C(=O)NC1=NC=CS1
It can be used as catalyst in pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be used as catalyst in pharmaceutical research.
Solubility
Solubility in water is almost transparent.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point >300°C
Quantity 5 g
Beilstein 3751377
Formula Weight 258.36
Percent Purity ≥98%
Assay (dry wt.), water <2%
Chemical Name or Material Sodium 1-undecanesulfonate