missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-dodecanesulfonate, 99%, Thermo Scientific Chemicals

Catalog Number p-7020765
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
2386-53-0
C12H25NaO3S
272.379
MFCD00007527
DAJSVUQLFFJUSX-UHFFFAOYSA-M
sodium 1-dodecanesulfonate, sodium dodecane-1-sulfonate, 1-dodecanesulfonic acid sodium salt, sodium laurylsulfonate, 1-dodecanesulfonic acid, sodium salt, unii-4gy1kjw8si, sodium dodecyl sulfonate, 4gy1kjw8si, laurylsulfonic acid sodium salt, 1-dodecanesulfonic acid, sodium salt 1:1
23665726
sodium;dodecane-1-sulfonate
CCCCCCCCCCCCS(=O)(=O)[O-].[Na+]
This item is not returnable. View return policy
2386-53-0
C12H25NaO3S
272.379
MFCD00007527
DAJSVUQLFFJUSX-UHFFFAOYSA-M
sodium 1-dodecanesulfonate, sodium dodecane-1-sulfonate, 1-dodecanesulfonic acid sodium salt, sodium laurylsulfonate, 1-dodecanesulfonic acid, sodium salt, unii-4gy1kjw8si, sodium dodecyl sulfonate, 4gy1kjw8si, laurylsulfonic acid sodium salt, 1-dodecanesulfonic acid, sodium salt 1:1
23665726
sodium;dodecane-1-sulfonate
CCCCCCCCCCCCS(=O)(=O)[O-].[Na+]

Sodium 1-dodecanesulfonate is used as a ion-associating reagent for HPLC for analysis of peptides and proteins. It acts as an ion-pair reagent, which increases the retention of thiabendazole on HPLC column during the determination of thiabendazole in fruits like citrus and banana. It is used in the synthesis of graphene-nanocomposites. It is also used as surfactant, flotation agent, foaming agent, penetrating agent and emulsifiers. It is involved in the preparation of 1-Chloro-dodecane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-dodecanesulfonate is used as a ion-associating reagent for HPLC for analysis of peptides and proteins. It acts as an ion-pair reagent, which increases the retention of thiabendazole on HPLC column during the determination of thiabendazole in fruits like citrus and banana. It is used in the synthesis of graphene-nanocomposites. It is also used as surfactant, flotation agent, foaming agent, penetrating agent and emulsifiers. It is involved in the preparation of 1-Chloro-dodecane.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point >300°C
Quantity 1 g
Beilstein 3919536
Solubility Information Soluble in water.
Formula Weight 272.39
Percent Purity 99%
Chemical Name or Material Sodium 1-dodecanesulfonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'