Learn More
Fisherbrand™ Soda Lime Glass Rimless Test Tube
Store test reagents and material. Fisherbrand™ soda lime glass rimless test tubes come in different sizes to meet your needs.
Brand: Fisherbrand™
93.55 DKK valid until 2024-04-30
Use promo code "21749" to get your promotional price.
Additional Details : Weight : 0.81900kg
Description
- Soda Lime Glass
- Rimless
- Disposable
Specifications
ISO 4142 | |
Soda Lime Glass | |
No |
100 mm | |
100 Pack | |
12 mm |