Få mere at vide
Tork Soap and Sanitiser Dispenser with Arm-Lever S4
The Tork Soap and Sanitizer Dispenser with Arm-lever is designed for use in areas where extra hygiene is critical and can be used with a wide range of Tork soaps, sanitizers and lotions in hygienically sealed bottles to suit your business needs.
886.60 DKK
Tekniske data
Produkttype | Soap Dispenser |
---|---|
Holder | Hand Soap, Hand Sanitizer |
Materiale | Plastic |
Montering | Wall Mount |
Antal rum | 1 |
Artikelnummer | Brand | Farve | Pris | Antal & tilgængelighed | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Artikelnummer | Brand | Farve | Pris | Antal & tilgængelighed | ||||||
30243698
|
Tork
564108 |
Black |
886.60 DKK
Hver |
Forventet forsendelse: 08-09-2025
Log ind for at se lagerbeholdningen. |
||||||
30243826
|
Tork
564100 |
White |
886.60 DKK
Hver |
Forventet forsendelse: 08-09-2025
Log ind for at se lagerbeholdningen. |
||||||
Beskrivelse
- Meet multiple hygiene needs with one system - wide range of soaps and sanitiser refills in foam, liquid or gel to suit your business and users' needs.
- Gentle-to-skin formulas - wide range of dermatologically tested, skin-friendly pH soaps, sanitisers and shower creams.
- Reliable dispensing - tested to last over one million pushes (based on durability test).
- Deliver the right dose - variable dose size options to optimise consumption, hygiene and the user experience.
Tekniske data
Soap Dispenser | |
Plastic | |
1 | |
Elevation |
Hand Soap, Hand Sanitizer | |
Wall Mount | |
Hinged Lid |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Produkttitel
Tork Soap and Sanitiser Dispenser with Arm-Lever S4